accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1304-28-5

IUPAC Name: oxobarium
Synonyms: barium oxide
Molecular Weight (g/mol): 153.33
Molecular Formula: BaO
InChi Key: QVQLCTNNEUAWMS-UHFFFAOYSA-N
SMILES: O=[Ba]
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)