Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Barium oxide

Catalog No. 5548475G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-484-75G 5 g
55-484-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-484-75G Supplier Sigma Aldrich Supplier No. 5548475G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Barium Oxide (BaO) is a white ionic crystal that has a non-flammable nature. It is a cubic structured compound that can be prepared by different methodologies, such as thermal decomposition of barium nitrate, and by heating barium carbonate with carbon black or tar.BaO can be used in a variety of applications such as:network modifier in the development of cathode ray tubesfabrication of glasses such as crown and borosilicate glassesstorage of nitrogen gaspreparation of biodiesel for internal combustion engines

Specifications

CAS 1304-28-5
Quantity 5 g
Molecular Formula BaO
Linear Formula BaO
MDL Number MFCD00003453
Synonym Baria; Barium monoxide
Molecular Weight (g/mol) 153.33
Percent Purity 99.99% trace metals basis
Density 5.72 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.