accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(E/Z)

CAS RN 13041-79-7

IUPAC Name: 4-(2-phenylethenyl)benzonitrile
Synonyms: 4-(2-phenylethenyl)benzonitrile
Molecular Weight (g/mol): 205.26
Molecular Formula: C15H11N
InChi Key: WQUHPLQCUQJSQW-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=CC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results