Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-trans-stilbene 96.0+%, TCI America™
SDP

Catalog No. C20461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20461G Supplier TCI America Supplier No. C20461G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13041-79-7
Molecular Formula C15H11N
Molecular Weight (g/mol) 205.26
MDL Number MFCD00489188
InChI Key WQUHPLQCUQJSQW-UHFFFAOYSA-N
Synonym trans-Stilbene-4-carbonitrile, (E)-4-Styrylbenzonitrile
PubChem CID 5375774
IUPAC Name 4-(2-phenylethenyl)benzonitrile
SMILES N#CC1=CC=C(C=CC2=CC=CC=C2)C=C1

Specifications

Melting Point 117°C
Quantity 1 g
UN Number 3439
Formula Weight 205.26
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-trans-stilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.