accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13054-98-3

IUPAC Name: 8-oxatricyclo[7.4.0.0²,⁷]tridecane
Synonyms: 8-oxatricyclo[7.4.0.0²,⁷]tridecane
Molecular Weight (g/mol): 180.29
Molecular Formula: C12H20O
InChi Key: XTDNMGZRUWMVLT-UHFFFAOYNA-N
SMILES: C1CCC2C(C1)OC1CCCCC21
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results