accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCl

CAS RN 13059-59-1

IUPAC Name: 8-chloro-8-boratricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene
Synonyms: 8-chloro-8-boratricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene
Molecular Weight (g/mol): 198.46
Molecular Formula: C12H8BCl
InChi Key: CWLYMXHRDXEBBF-UHFFFAOYSA-N
SMILES: ClB1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results