Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-5H-dibenzo[b,d]borole, TCI America™
SDP

Catalog No. C34211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C34211G Supplier TCI America Supplier No. C34211G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13059-59-1
Molecular Formula C12H8BCl
Molecular Weight (g/mol) 198.456
InChI Key CWLYMXHRDXEBBF-UHFFFAOYSA-N
Synonym 9-Chloro-9-borafluorene
PubChem CID 11241060
IUPAC Name 5-chlorobenzo[b][1]benzoborole
SMILES B1(C2=CC=CC=C2C3=CC=CC=C31)Cl

Specifications

Melting Point 52°C
Boiling Point 110°C
UN Number 1759
Quantity 1 g
Formula Weight 198.46
Percent Purity ≥95.0% (NMR)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-5H-dibenzo[b,d]borole
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.