accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1306-25-8

IUPAC Name: tellanylidenecadmium
Synonyms: cadmium telluride
Molecular Weight (g/mol): 240.01
Molecular Formula: CdTe
InChi Key: RPPBZEBXAAZZJH-UHFFFAOYSA-N
SMILES: [Cd]=[Te]
Physical Form 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)