Learn More
Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Cadmium telluride is used to make thin film solar cells. It can be alloyed with mercury to make a versatile infrared detector material (HgCdTe). It is used as an infrared optical material for optical windows and lenses and is proven to provide a good performance across a wide range of temperatures. It is also applied for electro-optic modulators and has the greatest electro-optic coefficient of the linear electro-optic effect among II-VI compound crystals. It can be doped with chlorine, which is used as a radiation detector for x-rays, gamma rays, beta particles and alpha particles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1041°C |
| Physical Form | Polycrystalline Pieces |
| Quantity | 25 g |
| Packaging | Glass bottle |
| Specific Gravity | 6.2 |
| Density | 6.2 |
| UN Number | UN3077 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 240 |
| Percent Purity | 99.999% |
| Show More |
Safety and Handling
| Hazard Category | H332-H350-H372 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P304+P340-P308+P313-P501c |
| EINECSNumber | 215-149-9 |
| RTECSNumber | EV3330000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.