accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1306-38-3

IUPAC Name: dioxocerium
Synonyms: cerium oxide
Molecular Weight (g/mol): 172.11
Molecular Formula: CeO2
InChi Key: CETPSERCERDGAM-UHFFFAOYSA-N
SMILES: O=[Ce]=O
Molecular Weight (g/mol) 
  • (8)
  • (1)
  • (31)
  • (3)
Quantity 
  • (2)
  • (2)
  • (13)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (10)
  • (1)
Physical Form 
  • (4)
  • (1)
  • (3)
  • (2)
  • (22)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (27)
  • (1)
  • (9)
  • (33)
  • (8)
  • (1)
  • (31)
  • (3)
  • (2)
  • (2)
  • (13)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (10)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (22)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)