accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNN

CAS RN 13063-43-9

IUPAC Name: dichloropropanedinitrile
Synonyms: propanedinitrile, dichloro-
Molecular Weight (g/mol): 134.95
Molecular Formula: C3Cl2N2
InChi Key: NCWGPYPDPWOJKC-UHFFFAOYSA-N
SMILES: ClC(Cl)(C#N)C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)