Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromalononitrile, 96%, Thermo Scientific™

Catalog No. AAB2177914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2177914 25 g
AAB2177903 1 g
AAB2177906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2177914 Supplier Thermo Scientific Chemicals Supplier No. B2177914

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13063-43-9
Molecular Formula C3Cl2N2
Molecular Weight (g/mol) 134.947
MDL Number MFCD00001841
InChI Key NCWGPYPDPWOJKC-UHFFFAOYSA-N
Synonym dichloromalononitrile, propanedinitrile, dichloro, dichlorodicyanomethane, malononitrile, dichloro, 2,2-dichloromalononitrile, dichloromalonitrile, propanedinitrile,2,2-dichloro, dichloropropanedinitrile, dichloro-malononitril, wln: ncxggcn
PubChem CID 25660
IUPAC Name 2,2-dichloropropanedinitrile
SMILES C(#N)C(C#N)(Cl)Cl

Specifications

Density 1.31
Boiling Point 96°C to 97°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.4335
Quantity 25 g
UN Number UN3276
Beilstein 906848
Formula Weight 134.95
Percent Purity 96%
Chemical Name or Material Dichloromalononitrile
Show More Show Less
Hazard Category H301+H311+H331-H315-H319
Hazard Statement GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 235-955-4
RTECSNumber OO3740000
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.