CAS RN 130674-54-3
CAS RN 130674-54-3
IUPAC Name:
(2R,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid
Synonyms:
(2R,3R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxybutanoic acid
Molecular Weight (g/mol):
341.36
Molecular Formula:
C19H19NO5
InChi Key:
OYULCCKKLJPNPU-PIGZYNQJSA-N
SMILES:
C[C@@H](O)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-D-Allo-Thr-OH, Advanced ChemTech
CAS: 130674-54-3 Molecular Formula: C19H19NO5 Molecular Weight (g/mol): 341.36 MDL Number: MFCD01318738 InChI Key: OYULCCKKLJPNPU-PIGZYNQJSA-N IUPAC Name: (2R,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid SMILES: C[C@@H](O)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O