accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13073-35-3

IUPAC Name: (2S)-2-amino-4-(ethylsulfanyl)butanoic acid
Synonyms: (.+-.)-ethionine ethionine (L)-ethionine L-ethionine ccris 289
Molecular Weight (g/mol): 163.24
Molecular Formula: C6H13NO2S
InChi Key: GGLZPLKKBSSKCX-YFKPBYRVSA-N
SMILES: CCSCC[C@H](N)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results