Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ethionine 98.0+%, TCI America™
SDP

Catalog No. E0039100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0039100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0039100MG Supplier TCI America Supplier No. E0039100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13073-35-3
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.235
MDL Number MFCD00002626
InChI Key GGLZPLKKBSSKCX-YFKPBYRVSA-N
PubChem CID 25674
ChEBI CHEBI:4886
IUPAC Name (2S)-2-amino-4-ethylsulfanylbutanoic acid
SMILES CCSCCC(C(=O)O)N

Specifications

Color White
Quantity 100 mg
Formula Weight 163.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Ethionine
RTECSNumber ES6825300
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.