CAS RN 13076-29-4
IUPAC Name:
1,4-dimethoxyanthracene
Synonyms:
1,4-dimethoxyanthracene
Molecular Weight (g/mol):
238.29
Molecular Formula:
C16H14O2
InChi Key:
YCTBFSAWJWMRGO-UHFFFAOYSA-N
SMILES:
COC1=C2C=C3C=CC=CC3=CC2=C(OC)C=C1
1
–
1
of
1
results
1
1,4-Dimethoxyanthracene 98%, Thermo Scientific™
CAS: 13076-29-4 Molecular Formula: C16H14O2 Molecular Weight (g/mol): 238.286 MDL Number: MFCD00051322 InChI Key: YCTBFSAWJWMRGO-UHFFFAOYSA-N Synonym: anthracene, 1,4-dimethoxy,anthracene,4-dimethoxy,d05gwi,anthracene,1,4-dimethoxy PubChem CID: 83107 IUPAC Name: 1,4-dimethoxyanthracene SMILES: COC1=CC=C(C2=CC3=CC=CC=C3C=C12)OC