Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dimethoxyanthracene 98%, Thermo Scientific™

Catalog No. AC407870050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407870050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407870050 Supplier Thermo Scientific Chemicals Supplier No. 407870050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13076-29-4
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00051322
InChI Key YCTBFSAWJWMRGO-UHFFFAOYSA-N
Synonym anthracene, 1,4-dimethoxy, anthracene,4-dimethoxy, d05gwi, anthracene,1,4-dimethoxy
PubChem CID 83107
IUPAC Name 1,4-dimethoxyanthracene
SMILES COC1=CC=C(C2=CC3=CC=CC=C3C=C12)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 133°C to 137°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Formula Weight 238.29
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Dimethoxyanthracene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.