accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3NHN

CAS RN 13078-13-2

IUPAC Name: 1-(3-methylphenyl)piperazine hydrochloridyl
Synonyms: 1-(3-methylphenyl)piperazine hydrochloridyl
Molecular Weight (g/mol): 211.71
Molecular Formula: C11H16ClN2
InChi Key: GLOZFOPZSXSVLZ-UHFFFAOYSA-N
SMILES: [Cl].CC1=CC=CC(=C1)N1CCNCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results