Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(m-Tolyl)piperazine Dihydrochloride 96.0+%, TCI America™
SDP

Catalog No. T131010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T131010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T131010G Supplier TCI America Supplier No. T131010G

Chemical Identifiers

CAS 13078-13-2
Molecular Formula C11H18Cl2N2
Molecular Weight (g/mol) 249.179
MDL Number MFCD00035284
InChI Key QJNWTYUDISMTMR-UHFFFAOYSA-N
PubChem CID 83110
IUPAC Name 1-(3-methylphenyl)piperazine;dihydrochloride
SMILES CC1=CC(=CC=C1)N2CCNCC2.Cl.Cl

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 249.18
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(m-Tolyl)piperazine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.