accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13080-90-5

IUPAC Name: bicyclo[2.2.1]hept-5-en-2-ol
Synonyms: 5-norbornen-2-ol (exo)
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: MKOSBHNWXFSHSW-UHFFFAOYNA-N
SMILES: OC1CC2CC1C=C2
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results