Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Norbornen-2-ol, mixture of endo and exo

Catalog No. N3210525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N3210525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N3210525G Supplier Sigma Aldrich Supplier No. N3210525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Geranylamine is an di-isoprene derivative.geranylamine may be employed as starting reagent for the total synthesis of a naturally occurring guanidine alkaloid, nitensidine d
  • It may be used for the synthesis of 5-alkylamino- and 2,5-bis(alkylamino)-[1,4]-benzoquinone

Specifications

CAS 13080-90-5
Melting Point 104°C to 106°C (lit.)
Molecular Formula C7H10O
Linear Formula C7H10O
MDL Number MFCD00167566
Quantity 5 g
Molecular Weight (g/mol) 110.15
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.