accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOCH3H3CCH3H2NHCl

CAS RN 13081-32-8

IUPAC Name: tert-butyl 2-amino-4-methylpentanoate hydrochloride
Synonyms: tert-butyl 2-amino-4-methylpentanoate hydrochloride
Molecular Weight (g/mol): 223.74
Molecular Formula: C10H22ClNO2
InChi Key: RFUWRXIYTQGFGA-UHFFFAOYNA-N
SMILES: Cl.CC(C)CC(N)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)