Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Leucine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. L01965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01965G 5 g
L01961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01965G Supplier TCI America Supplier No. L01965G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13081-32-8
Molecular Formula C10H22ClNO2
Molecular Weight (g/mol) 223.741
MDL Number MFCD00083642
InChI Key RFUWRXIYTQGFGA-DDWIOCJRSA-N
Synonym h-d-leu-otbu.hcl, r-tert-butyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-otbu hcl, d-leucine tert-butyl ester hydrochloride, d-leucine t-butyl ester hydrochloride, tert-butyl 2r-2-amino-4-methylpentanoate hydrochloride, r-tert-butyl 2-amino-4-methylpentanoate hcl, h-d-leu-otbu. hcl, d-leucine tert.butyl ester hydrochloride, d-leucine-t-butylester hydrochloride
PubChem CID 44629930
IUPAC Name tert-butyl (2R)-2-amino-4-methylpentanoate;hydrochloride
SMILES CC(C)CC(C(=O)OC(C)(C)C)N.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 223.74
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Leucine tert-Butyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.