accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSiClClSiClClCl

CAS RN 13083-94-8

IUPAC Name: trichloro[6-(trichlorosilyl)hexyl]silane
Synonyms: trichloro[6-(trichlorosilyl)hexyl]silane
Molecular Weight (g/mol): 353.03
Molecular Formula: C6H12Cl6Si2
InChi Key: ICJGKYTXBRDUMV-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CCCCCC[Si](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results