Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,6-Bis(trichlorosilyl)hexane

Catalog No. 45224610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-224-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-224-610G Supplier Sigma Aldrich Supplier No. 45224610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,6-Bis(trichlorosilyl)hexane is a silane based cross-linking agent that can be used in the synthesis of polymeric blended films. These films are majorly utilized as dielectrics for applications in organic electronic based devices such as organic field effect transistors (OFETs) and organic thin film transistors (OTFTs).

Specifications

CAS 13083-94-8
Density 1.327 g/mL (at 25°C)
Boiling Point 281°C
Molecular Formula C6H12Cl6Si2
Refractive Index n20/D 1.475 (literature)
Linear Formula Cl3Si(CH2)6 SiCl3
MDL Number MFCD00053189
Quantity 10 g
Synonym 1,6-Bis(trichlorosilyl)hexane
Molecular Weight (g/mol) 353.05
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.