accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1308671-90-0

IUPAC Name: 3-(5-bromothiophen-2-yl)-2,5-bis(2-ethylhexyl)-6-(thiophen-2-yl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Synonyms: 3-(5-bromothiophen-2-yl)-2,5-bis(2-ethylhexyl)-6-(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4-dione
Molecular Weight (g/mol): 603.68
Molecular Formula: C30H39BrN2O2S2
InChi Key: IUJFPKGBQVGKRW-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C(=O)C2=C(N(CC(CC)CCCC)C(=O)C2=C1C1=CC=CS1)C1=CC=C(Br)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results