Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(5-Bromo-2-thienyl)-2,5-bis(2-ethylhexyl)-6-(2-thienyl)pyrrolo[3,4-c]pyrrole-1,4-dione 98.0+%, TCI America™
SDP

Catalog No. B5456200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5456200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5456200MG Supplier TCI America Supplier No. B5456200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1308671-90-0
Molecular Formula C30H39BrN2O2S2
Molecular Weight (g/mol) 603.678
MDL Number MFCD26403655
InChI Key IUJFPKGBQVGKRW-UHFFFAOYSA-N
PubChem CID 59629052
IUPAC Name 4-(5-bromothiophen-2-yl)-2,5-bis(2-ethylhexyl)-1-thiophen-2-ylpyrrolo[3,4-c]pyrrole-3,6-dione
SMILES CCCCC(CC)CN1C(=C2C(=C(N(C2=O)CC(CC)CCCC)C3=CC=C(S3)Br)C1=O)C4=CC=CS4

Specifications

Melting Point 126°C
Quantity 200 mg
Formula Weight 603.68
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(5-Bromo-2-thienyl)-2,5-bis(2-ethylhexyl)-6-(2-thienyl)pyrrolo[3,4-c]pyrrole-1,4-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.