accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CCH3CH3OH

CAS RN 1309980-11-7

IUPAC Name: 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
Synonyms: 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
Molecular Weight (g/mol): 262.16
Molecular Formula: C15H23BO3
InChi Key: NNPQWBXXZHNLAW-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results