Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Hydroxy-2-propanyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AC445070050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445070050 Supplier Thermo Scientific Chemicals Supplier No. 445070050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309980-11-7
InChI Key NNPQWBXXZHNLAW-UHFFFAOYSA-N
Synonym 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol, 3-2-hydroxypropan-2-yl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propyl phenylboronic acid pinacol ester, 3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester, 3-2-hydroxypropan-2-yl phenyl boronic acid pinacol ester, 2-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl propan-2-ol, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol
PubChem CID 46738028
IUPAC Name 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(C)(C)O

Specifications

Assay Percent Range 97%
Molecular Formula C15H23BO3
Quantity 5 g
Molecular Weight (g/mol) 262.156
Formula Weight 262.16
Chemical Name or Material 3-(2-Hydroxy-2-propanyl)phenylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.