accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131-22-6

IUPAC Name: 4-(2-phenyldiazen-1-yl)naphthalen-1-amine
Synonyms: 4-(2-phenyldiazen-1-yl)naphthalen-1-amine
Molecular Weight (g/mol): 247.3
Molecular Formula: C16H13N3
InChi Key: IICHURGZQPGTRD-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results