Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylazo-1-naphthylamine 95.0+%, TCI America™
SDP

Catalog No. P058425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P058425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P058425G Supplier TCI America Supplier No. P058425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131-22-6
Molecular Formula C16H13N3
Molecular Weight (g/mol) 247.30
MDL Number MFCD00004025
InChI Key IICHURGZQPGTRD-UHFFFAOYSA-N
Synonym Naphthyl Red, Solvent Yellow 4
PubChem CID 8562
IUPAC Name 4-(2-phenyldiazen-1-yl)naphthalen-1-amine
SMILES NC1=C2C=CC=CC2=C(C=C1)N=NC1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 247.30
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylazo-1-naphthylamine
EINECSNumber (4)-0335
RTECSNumber QJ0360000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.