accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOO

CAS RN 131-70-4

IUPAC Name: 2-(butoxycarbonyl)benzoic acid
Synonyms: monobutyl phthalate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: YZBOVSFWWNVKRJ-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=CC=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results