Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobutyl Phthalate 95.0+%, TCI America™
SDP

Catalog No. P11325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11325G 5 g
P113225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11325G Supplier TCI America Supplier No. P11325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131-70-4
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00043498
InChI Key YZBOVSFWWNVKRJ-UHFFFAOYSA-N
Synonym Phthalic Acid Monobutyl Ester, Butyl Hydrogen Phthalate
PubChem CID 8575
IUPAC Name 2-(butoxycarbonyl)benzoic acid
SMILES CCCCOC(=O)C1=CC=CC=C1C(O)=O

Specifications

Melting Point 73°C
Color White
Quantity 5 g
Formula Weight 222.24
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monobutyl Phthalate
EINECSNumber (3)-1308
RTECSNumber TI2475000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.