accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiH2OOH

CAS RN 1310-66-3

IUPAC Name: lithium(1+) hydrate hydroxide
Synonyms: lithium hydroxide monohydrate
Molecular Weight (g/mol): 41.96
Molecular Formula: H3LiO2
InChi Key: GLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES: [Li+].O.[OH-]
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (9)
  • (1)
Percent Purity 
  • (6)
  • (2)
  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (2)
  • (8)
  • (5)
Grade 
  • (9)
  • (3)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (15)
  • (4)
  • (1)
  • (5)
  • (2)
  • (4)
  • (4)
  • (20)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (9)
  • (1)
  • (6)
  • (2)
  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (8)
  • (5)
  • (9)
  • (3)
  • (3)
  • (1)
  • (2)