Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hydroxide monohydrate, Solstice

Catalog No. 6000903 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-009-03 500 g
60-009-01 25 g
60-009-02 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-009-03 Supplier Solstice Supplier No. 402974500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
IUPAC Name lithium;hydroxide;hydrate
SMILES [Li+].O.[OH-]

Specifications

Quantity 500 g
Assay Percent Range ≥98.0%
Linear Formula LiOH · H2O
UN Number UN2680
Solubility Information 190 to 220 g/L
Formula Weight 41.96g/mol
Percent Purity ≥98%
Odor Odorless
Packaging Plastic Bottle
Sensitivity Air Sensitive
Chemical Name or Material Lithium hydroxide monohydrate
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.