accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13103-75-8

IUPAC Name: N,N-dimethyl-4-[2-(pyridin-2-yl)diazen-1-yl]aniline
Synonyms: N,N-dimethyl-4-[2-(pyridin-2-yl)diazen-1-yl]aniline
Molecular Weight (g/mol): 226.28
Molecular Formula: C13H14N4
InChi Key: IOJNPSPGHUEJAQ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results