Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-2-azo-p-dimethylaniline 98.0+%, TCI America™
SDP

Catalog No. P06171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06171G Supplier TCI America Supplier No. P06171G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13103-75-8
Molecular Formula C13H14N4
Molecular Weight (g/mol) 226.283
MDL Number MFCD00010435
InChI Key IOJNPSPGHUEJAQ-UHFFFAOYSA-N
Synonym 4-2-pyridylazo-n,n-dimethylaniline, pada, pyridine-2-azo-p-dimethylaniline, pyridine-2-azodimethylaniline, n,n-dimethyl-4-2-pyridylazo aniline, 2-4-dimethylaminophenylazo pyridine, p-2-pyridylazo dimethylaniline, 2-p-n,n-dimethylaminophenylazo pyridine, n,n-dimethyl-4-pyridin-2-yldiazenyl aniline, pyridine, 2-p-dimethylamino phenyl azo
PubChem CID 81528
IUPAC Name N,N-dimethyl-4-(pyridin-2-yldiazenyl)aniline
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=N2

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 226.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2-azo-p-dimethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.