accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131083-16-4

IUPAC Name: 5,11-diethyl-2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 5,11-diethyl-2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 318.22
Molecular Formula: C18H25BF2N2
InChi Key: DZSMVBDAUBBZJD-UHFFFAOYSA-N
SMILES: CCC1=C(C)N2C(=C1C)C(C)=C1C(C)=C(CC)C(C)=[N+]1[B-]2(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results