Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrromethene 567 98.0+%, TCI America™
SDP

Catalog No. E1065200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1065200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E1065200MG Supplier TCI America Supplier No. E1065200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 131083-16-4
Molecular Formula C18H25BF2N2
Molecular Weight (g/mol) 318.219
InChI Key DZSMVBDAUBBZJD-UHFFFAOYSA-N
Synonym Pyrromethene 567
PubChem CID 14766994
SMILES [B-]1(N2C(=C(C(=C2C(=C3[N+]1=C(C(=C3C)CC)C)C)C)CC)C)(F)F

Specifications

Melting Point 213°C
Quantity 200 mg
Formula Weight 318.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [[(4-Ethyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](dif
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.