accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1311933-83-1

OOHOONHH3CCH2

CAS RN 1311933-83-1

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylnon-8-enoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylnon-8-enoic acid
Molecular Weight (g/mol): 407.51
Molecular Formula: C25H29NO4
InChi Key: WSTJGDVRPQAMSC-VWLOTQADSA-N
SMILES: CC(CCCCCC=C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)