CAS RN 1312-81-8
Lanthanum(III) oxide, 99.99%, (trace metal basis)
CAS: 1312-81-8 Molecular Formula: La2O3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011071
Lanthanum(III) oxide, 99.9%, (trace metal basis)
CAS: 1312-81-8 Molecular Formula: La2O3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011071
Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade
CAS: 1312-81-8 Molecular Formula: La2O3 MDL Number: MFCD00011071
Lanthanum(III) oxide, REacton™, 99.99% (REO)
CAS: 1312-81-8 Molecular Formula: La2O3 MDL Number: MFCD00011071
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
CAS: 1312-81-8 Molecular Formula: La2O3 in 5% HNO3 MDL Number: MFCD00011066
Lanthanum(III) oxide, 91%
CAS: 1312-81-8 Molecular Formula: La2O3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011071
Lanthanum(III) Oxide, MilliporeSigma™
CAS: 1312-81-8 Molecular Formula: La2O3 Synonym: Lanthanum III Oxide
Lanthanum Oxide, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 1312-81-8 Molecular Formula: La2O3
Lanthanum(III) oxide, REacton™, 99.999% (REO)
CAS: 1312-81-8 Molecular Formula: La2O3 MDL Number: MFCD00011071
Lanthanum(III) oxide, REacton™, 99.9% (REO)
CAS: 1312-81-8 MDL Number: MFCD00011071 Synonym: Lanthanum trioxide,Lanthanum sesquioxide
Lanthanum(III) oxide, For AAS, ≥99.9%, MilliporeSigma™ Supelco™
MDL Number: MFCD00011071
Lanthanum Oxide, 99.99%, MP Biomedicals
CAS: 1312-81-8 Molecular Weight (g/mol): 325.82 g/moL
Lanthanum(III) oxide, 99.9% (metals basis)
CAS: 1312-81-8 MDL Number: MFCD00011071
Lanthanum(III) Oxide 99.9+%, TCI America™
CAS: 1312-81-8 Molecular Formula: La2O3 MDL Number: MFCD00011071
- 1
- 2