Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) oxide, For AAS, ≥99.9%, MilliporeSigma™ Supelco™

Catalog No. 111012930 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2930 50 g
11-101-2929 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2930 Supplier MilliporeSigma Supelco Supplier No. 28920550G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lanthanum (III) oxide is an alkaline earth oxide, and is a diamagnetic, isostructural analog of neodymium oxide. It is also an efficient recyclable catalyst, and can be used in coupling various aryl halides with aromatic/alkyl thiols.

  • AAS grade trace metals basis reagent
  • Recyclable catalyst for coupling reactions
  • Precursor to LAMOX fast ion conductors and superconductors
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011071

Specifications

CAS 1312-81-8
Physical Form Powder
Quantity 50 g
Assay Percent Range ≥99.9%
Molecular Formula La2O3
Linear Formula La2O3
Solubility Information Insoluble in Water
InChI Formula InChI=1S/2La.3O
InChI Key KTUFCUMIWABKDW-UHFFFAOYSA-N
Formula Weight 325.81
Percent Purity ≥99.9% (trace metals basis); ≥99.9%
Grade AAS
Packaging Poly Bottle
Appearance White Powder
Specific Gravity 6.51 at 25°C
Density 6.51 g/mL (at 25°C)
Assay Trace Metals Basis
Show More Show Less
RTECSNumber OE5330000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.