accessibility menu, dialog, popup

UserName

Viewing profile as user:
PH

CAS RN 131211-27-3

IUPAC Name: bis(adamantan-1-yl)phosphane
Synonyms: bis(adamantan-1-yl)phosphane
Molecular Weight (g/mol): 302.44
Molecular Formula: C20H31P
InChi Key: RRRZOLBZYZWQBZ-UHFFFAOYSA-N
SMILES: C1C2CC3CC1CC(C2)(C3)PC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)