Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-1-Adamantylphosphine

Catalog No. 695823500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-582-3500MG 500 mg
69-582-32G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-582-3500MG Supplier Sigma Aldrich Supplier No. 695823500MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Di-1-Adamantylphosphine

Specifications

CAS 131211-27-3
Melting Point 181°C to 185°C
Molecular Formula C20H31P
Linear Formula C20H31P
MDL Number MFCD23701602
Quantity 500 mg
Synonym Bis(adamant-1-yl)phosphine; Bis(tricyclo[3.3.1.13,7]dec-1-yl)phosphine
Molecular Weight (g/mol) 302.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.