accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13125-66-1

IUPAC Name: bromo(heptyl)magnesium
Synonyms: bromo(heptyl)magnesium
Molecular Weight (g/mol): 203.41
Molecular Formula: C7H15BrMg
InChi Key: LDKGBIJDNGSVIH-UHFFFAOYSA-M
SMILES: CCCCCCC[Mg]Br
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)