Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™

Catalog No. AC433031000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433031000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433031000 Supplier Thermo Scientific Chemicals Supplier No. 433031000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13125-66-1 | C7H15BrMg | 203.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13125-66-1 , 60-29-7
Molecular Formula C7H15BrMg
Molecular Weight (g/mol) 203.41
MDL Number MFCD00013517
InChI Key LDKGBIJDNGSVIH-UHFFFAOYSA-M
Synonym n-heptylmagnesium bromide, 1-heptylmagnesium bromide, heptyl magnesium bromide, magnesium, bromoheptyl, n-heptyl magnesium bromide, heptylmagnesium bromide solution, ldkgbijdngsvih-uhfffaoysa-m, 1-heptylmagnesium bromide 0.25 m in tetrahydrofuran, heptylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
PubChem CID 10910616
IUPAC Name magnesium;heptane;bromide
SMILES CCCCCCC[Mg]Br

Specifications

Color Brown
Density 0.8410g/mL
Boiling Point 35.0°C
Flash Point −30°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)6MgBr
Quantity 100 mL
Specific Gravity 0.841
Solubility Information Solubility in water: reacts
Formula Weight 203.4
Physical Form Liquid
Chemical Name or Material Heptylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Extremely flammable liquid and vapor.
Reacts violently with water.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/Bond container and receiving equipment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mout

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.