accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHNH2(E/Z)

CAS RN 1312766-67-8

IUPAC Name: N'-hydroxy-2-(4-methylphenyl)ethanimidamide
Synonyms: N'-hydroxy-2-(4-methylphenyl)ethanimidamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: CKMSCSXWLFGEGO-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC(N)=NO)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results