accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CH3CCH3PHCH3CH3H3CCH3CH3H3C

CAS RN 131322-08-2

IUPAC Name: tetraphenylboranuide; tri-tert-butylphosphanium
Synonyms: tri-tert-butylphosphanium tetraphenylborate
Molecular Weight (g/mol): 522.56
Molecular Formula: C36H48BP
InChi Key: QWISVPBFGJWCBS-UHFFFAOYSA-O
SMILES: CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results