Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-tert-butylphosphonium Tetraphenylborate 98.0+%, TCI America™
SDP

Catalog No. T22581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22581G Supplier TCI America Supplier No. T22581G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131322-08-2
Molecular Formula C36H48BP
Molecular Weight (g/mol) 522.564
InChI Key QWISVPBFGJWCBS-UHFFFAOYSA-O
PubChem CID 23136663
IUPAC Name tetraphenylboranuide;tritert-butylphosphanium
SMILES [B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C

Specifications

Color White
Quantity 1 g
UN Number 2811
Formula Weight 522.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tri-tert-butylphosphonium Tetraphenylborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.