accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131379-39-0

IUPAC Name: 4-ethoxy-4-oxobutan-1-ide; bromozincylium
Synonyms: 4-ethoxy-4-oxobutan-1-ide; bromozincylium
Molecular Weight (g/mol): 260.44
Molecular Formula: C6H11BrO2Zn
InChi Key: XPARCVGSTPKNNR-UHFFFAOYSA-M
SMILES: [Zn+]Br.CCOC(=O)CC[CH2-]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)